missing translation for 'onlineSavingsMsg'
Learn More

Merck Ultrafiltration Membranes - YM

High-recovery membranes for stirred cells

Brand:  Merck 14432

Code : 72

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10722551

  • £625.00 / Pack of 10
Estimated Shipment: 17-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Adsorptive
100,000 NMWL
28.7 cm2
10 Pack
2.5 in.
Regenerated Cellulose
YM-100
Protein purification, protein concentration, protein desalting
Ultrafiltration Disc
6.35 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers