Learn More
EMD Millipore™ Ultrafiltration Membranes - YM
High-recovery membranes for stirred cells
Brand:  EMD Millipore™ 13651
Code : 72
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10440201
  • £241.00 / Pack of 5
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications
Specifications
9cm
Adsorptive
Regenerated Cellulose
Protein purification, protein concentration, protein desalting
5 Pack
YM-10
63cm2
10000 NMWL
Ultrafiltration Disc
Videos
Documentation
Technical Documents