Learn More
EMD Millipore™ Ultrafiltration Membranes - Amicon™ YC Cellulose Acetate Filters
High-recovery membranes for stirred cells
Brand:  EMD Millipore™ 13051
Code : NEW
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10064350
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Specifications
9cm
6300cm2
500 NMWL
Ultrafiltration Membrane
Ultrafiltration
Cellulose Acetate
Concentration of hydrophobic samples or to desalt extremely dilute solutions for stirred cells
1 Pack
Videos