Learn More
EMD Millipore Ultrafiltration Membranes - Amicon™ XM Polyacrylonitrile/PVC Copolymer Filters
High-recovery membranes for stirred cells
Brand:  EMD Millipore 14332
UNSPSC : 40161507
Code : NEW
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10126570
  • £336.90 / Pack of 10
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications Description & Specifications
Specifications
6.2cm
300000 NMWL
Ultrafiltration Disc
Polyacrylonitrile/PVC Co-polymer
High recovery membranes for stirred cells
10 Pack