missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Ultrafiltration Membranes - Amicon™ XM Polyacrylonitrile/PVC Copolymer Filters

High-recovery membranes for stirred cells

Brand:  EMD Millipore™ 14322

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10557282

  • £260.00 / Pack of 10


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Regenerated cellulose membranes
  • For concentration of hydrophobic samples or to desalt extremely dilute solutions
  • Hydrophilic, tight microstructure assures high retention with low adsorption of protein, DNA, or other macromolecules

†43mm nominal diameter.
‡62mm nominal diameter.

Specifications

Specifications

4.45cm
300000 NMWL
Ultrafiltration Disc
Polyacrylonitrile/PVC Co-polymer
High recovery membranes for stirred cells
10 Pack
Videos