Learn More
EMD Millipore™ Ultrafiltration Membranes - Amicon™ XM Polyacrylonitrile/PVC Copolymer Filters
High-recovery membranes for stirred cells
Brand:  EMD Millipore™ 14132AM
Code : NEW
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10491731
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Specifications
6.35cm
50000 NMWL
Ultrafiltration Disc
Polyacrylonitrile/PVC Co-polymer
High recovery membranes for stirred cells
10 Pack
Videos