Learn More
EMD Millipore™ Ultrafiltration Membranes - Amicon™ XM Polyacrylonitrile/PVC Copolymer Filters
High-recovery membranes for stirred cells
Brand:  EMD Millipore™ 14112AM
Code : NEW
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10218380
  • £149.00 / Pack of 10
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Regenerated cellulose membranes
  • For concentration of hydrophobic samples or to desalt extremely dilute solutions
  • Hydrophilic, tight microstructure assures high retention with low adsorption of protein, DNA, or other macromolecules
Specifications
Specifications
2.5cm
50000 NMWL
Ultrafiltration Disc
Polyacrylonitrile/PVC Co-polymer
High recovery membranes for stirred cells
10 Pack
Videos