missing translation for 'onlineSavingsMsg'
Learn More

Merck Ultrafiltration Discs, 100 kDa NMW, Ultracel™ Regenerated Cellulose

100 kDa NMWL, Ultracel™ regenerated cellulose

Brand:  Merck PLHK09005

Code : 72

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10536722

  • £400.00 / Pack of 5
Estimated Shipment: 20-06-2024
to see stock.
Add to basket

Description

Description

100 kDa NMWL, Ultracel™ regenerated cellulose

Assemble your Amicon Stirred Cell with the membrane of your choice. Ultrafiltration cut disc membranes are available in Ultracel™ regenerated cellulose and Biomax™ polyethersulvone (PES). The membranes are available in a range of different pore sizes & diameters.Ultracel regenerated cellulose membranes are recommended for concentrating or desalting dilute solutions. The hydrophilic, tight microstructure of Ultracel membranes assures the highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules.Biomax PES membranes are recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media.Microfiltration Durapore™ PVDF cut discs are available 0.1 μm, 0.22 μm, and 0.45 μm pore sizes in both 63.5 mm and 75 mm diameters.Features & Benefits:•Disc diameters include: 25, 44.5, 47, 63.5, 76, 90 and 150 mm •Both Ultracel™, Durapore™, and Biomax™ membranes are available in a wide range of pore sizes to meet your separation requirements •Compatible with Amicon™ Stirred Cells Applications:Concentration, Buffer Exchange, Desalting
Specifications

Specifications

Ultrafiltration
100 kDa
63 cm2
Regenerated Cellulose (RC)
Ultrafiltration Disc
Regenerated Cellulose
PLHK
Protein purification; protein concentration; protein desalting
5Pack
9 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers