Learn More
Merck Millex™-AA Syringe Filter Unit, 0.80 μm
A 25 mm diameter sterile syringe filter with a 0.8 μm pore size Mixed Cellulose Esters membrane.
Brand: Merck SLAAV255F
Code : 71
Additional Details : Weight : 0.10000kg
Description
Millex™ sterile syringe filters are ideal for sterilizing organic solvents, aqueous solutions or air/gas. Many sample preparation methods specify Millex™ filters because of their unsurpassed quality and consistency. Millex™ filters not only feature minimal hold-up volume and faster flow rates, but they also withstand higher operating pressures and are chemically compatible with a range of samples and solvents to minimize leaching of extractable impurities. Millex™ filters are produced in a controlled, automated environment with the highest quality standards. Sterile devices come with a certificate of quality, with select devices marked for medical applications.Features & Benefits:
- Tailored to a variety of applications based on membrane type, housing and validation rating
- Medical Device-rated devices in some countries including vented devices for direct patient care
- Specialized filter units to protect hemodialysis transducers from blood and moisture and for vacuum line protection
- Low extractables and low absorption membranes for minimum sample losses
- Faster flow rates and higher operating pressures for high performance needs
Applications:
Sterilization of Aqueous Solutions, Organic Solvents and Gases, Filtration of High Value Protein or Small Molecule Components, Select Medical Device Applications (Some Including Direct Patient Care Depending on Country/Region)
Specifications
Not Autoclavable | |
0.8 μm | |
Clarification of water, buffers or aqueous solutions | |
500 mL |
Single-use | |
Sterile | |
Mixed Cellulose Esters (MCE) | |
2.5 cm |