missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Sterifil™ 47mm Aseptic Vacuum Filter System Replacement Parts

Used for general filtration applications and filtration of samples for particulate or biological contamination analysis

Brand:  EMD Millipore™ XX1104704

Code : 71

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10756711

  • £25.75 / Each


Estimated Shipment
13-06-2018
Add to basket

Specifications

Specifications

250mL Replacement Funnel
Millipore Sterifil 47mm Aseptic Vacuum Filter System
250mL
Videos