missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Sterifil™ 47mm Aseptic Vacuum Filter System Replacement Parts

Used for general filtration applications and filtration of samples for particulate or biological contamination analysis

Brand:  EMD Millipore™ XX11J4750

Code : 72

Additional Details : Weight : 1.70100kg

 View more versions of this product

Product Code. 10737671

  • £114.00 / Each


Estimated Shipment
20-06-2018
Add to basket

Specifications

Specifications

500mL Sterifil Holder
Millipore Sterifil 47mm Aseptic Vacuum Filter System
13.8cm2
Polysulfone, Polypropylene, Silicone
500mL
Videos