missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Replacement PTFE O-Ring for Stainless-Steel 142mm Pressure Filter Holder
PTFE O-Ring
Brand: Merck YY2214253
70.95 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Code : 72
Additional Details : Weight : 0.10000kg
Description
- Replacement part for Stainless Steel 142mm Filter Holders
- Compatible for use with YT30142HW Hazardous Waste Filtration System
- Compatible for use with YY3014236 (142mm) Stainless Steel Filter Holders
- PTFE construction is inert and chemically resistant
Specifications
Not Autoclavable | |
PTFE | |
O-Ring, 2-251 |
142 mm Stainless Steel Filter Holder (YY3014236) | |
Non-sterile |