EMD Millipore SNAP i.d.™ Protein Detection System: Blot Holders
Blot holders for use with EMD Millipore SNAP i.d. Protein Detection System
Overview
Brand: EMD Millipore
Manufacturer Part Number: WBAVDBH02
X30 Double blot holder SNAP i.d. for WesternStück
UNSPSC: 41105300
Code: 50
Additional Details:
Additional Details: Weight: 2.49480kg
Product Code. 10508294
Quantity Price
1 £ 308.7 / Pack of 30
Estimated Shipment
31-03-2017
Add to basket
  • Description and Specification
  • Documentation
  • Accessories and Spare Parts
Description and Specification
Specification
For Use With (Equipment) Snap i.d.™ Protein detection system
Quantity 30/Pk.
Product Type Double Well Blot Holder
Device Holds 4.6 x 8.8cm Blots
No. of Wells 2
Includes Spacers
For Use With (Application) Western Blotting
Documentation Accessories and Spare Parts