Learn More
Merck SNAP i.d. 2.0™ IHC Slide Holders
For use with SNAP i.d. 2.0 protein detection system
Brand: Merck SNAP2SH
Additional Details : Weight : 0.10000kg
Description
- Disposable slide holders fit standard 25 x 75 x 1mm glass slide
- Each slide holder features antibody injection/recovery ports to facilitate addition and removal of small volumes of antibodies and reagents
- Specially designed gaskets eliminate need for pap pens
- For processing 24 tissue slides
Specifications
24/Pk. |