missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Scepter 2.0 Automated Handheld Cytometer

Cell counting in a portable, personalized format

Brand:  EMD Millipore™ PHCC20040

Code : 72

Additional Details : Weight : 0.01250kg

 View more versions of this product

Product Code. 11359791

  • £3275.33 / Each


Estimated Shipment
13-06-2018
Add to basket

Description

Description

Scepter Hand-Held Cytometer

  • Automated, impedance-based counting without the error that accompanies vision-based systems
  • Accurately count cells in the 4-25μm diameter range
  • In addition to cell concentration, the Scepter screen displays a histogram showing the size or volume distribution of the cell population
  • On-screen instructions will walk user through assay, step by step
  • In less than 20 seconds (on average) the Scepter cytometer will display all the data needed regarding the culture
  • After the count is complete, use the toggle button to move from one histogram to another, manipulate, or gate counts
  • See into the health and quality of cell culture from one preparation to the next
Scepter Sensor
  • Has a precision drilled aperture through which cells pass and are counted based on the Coulter principle of impedance changes across a current established by sensor electrodes
  • Ensures reproducible counts by drawing precisely 50μL of sample into its liquid handling microchannel each time
  • Eliminates pipetting errors and uneven sample loading that can occur
  • Able to count cells between 4 and 25μm in diameter, as both 60μm- and 40μm-aperture sensors are available
Scepter Software Pro
  • Compare several samples and data sets side by side using histogram overlay and multiparametric tables
  • Save and create gating methods to be used from one experiment to the next
  • Create attractive graphical presentations and reports with data

Specifications

Specifications

<30 sec.
40μm sensors (50 pack); Test bead vial, USB cable, software, 2 O-rings
10,000 to 500,000
50 Pk.
Automated
Counting cells and delivering histrogram profiles of cellular populations
4 to 25μm
Stores up to 72 counts
6 to 36μm
CHO cells, HeLa cells and mouse embryonic stem cells, can even be used to count subpopulations in peripheral Blood mononuclear cell preps
100μL
Automated Cell Counter
Videos
Documentation View accessory products