missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Scepter 2.0 Automated Handheld Cytometer

Cell counting in a portable, personalized format

Brand:  EMD Millipore™ PHCC20060

Code : 72

Additional Details : Weight : 0.99790kg

 View more versions of this product

Product Code. 11369791

  • £3985.00 / Each


Estimated Shipment
09-05-2018
Add to basket

Description

Description

Scepter Hand-Held Cytometer

  • Automated, impedance-based counting without the error that accompanies vision-based systems
  • Accurately count cells in the 4-25μm diameter range
  • In addition to cell concentration, the Scepter screen displays a histogram showing the size or volume distribution of the cell population
  • On-screen instructions will walk user through assay, step by step
  • In less than 20 seconds (on average) the Scepter cytometer will display all the data needed regarding the culture
  • After the count is complete, use the toggle button to move from one histogram to another, manipulate, or gate counts
  • See into the health and quality of cell culture from one preparation to the next
Scepter Sensor
  • Has a precision drilled aperture through which cells pass and are counted based on the Coulter principle of impedance changes across a current established by sensor electrodes
  • Ensures reproducible counts by drawing precisely 50μL of sample into its liquid handling microchannel each time
  • Eliminates pipetting errors and uneven sample loading that can occur
  • Able to count cells between 4 and 25μm in diameter, as both 60μm- and 40μm-aperture sensors are available
Scepter Software Pro
  • Compare several samples and data sets side by side using histogram overlay and multiparametric tables
  • Save and create gating methods to be used from one experiment to the next
  • Create attractive graphical presentations and reports with data

Specifications

Specifications

<30 sec.
60μm sensors (50 pack); Test bead vial, USB cable, software, 2 O-rings
10,000 to 500,000
50 Pk.
Automated
Counting cells and delivering histrogram profiles of cellular populations
4 to 25μm
Stores up to 72 counts
6 to 36μm
CHO cells, HeLa cells and mouse embryonic stem cells, can even be used to count subpopulations in peripheral Blood mononuclear cell preps
100μL
Automated Cell Counter
Videos
Documentation