Learn More
Merck Millipore RoClean™ Cleaning Pouches for Water Purification Systems
Keep your purification system clean with easy-to-use RoClean pouches.
£278.00
Specifications
For Use With (Equipment) Elix™, Milli-RO™, Milli-RX™ and Millipore™ RiOs™ water purification system
View More Specs
Products 4
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
10040272 View Documents Merck Millipore
ZWACID012
Acid cleaning pouch £278.00
Pack of 12 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Add to basket
12048812 View Documents Merck Millipore
ZWACID012
Acid cleaning pouch £278.00
Pack of 12 View alternative products
Save Up to 
»
Estimated Shipment
17-03-2020
Add to basket
12058812 View Documents Merck Millipore
ZWBASE012
Base cleaning pouch £278.00
Pack of 12 View alternative products
Save Up to 
»
Estimated Shipment
18-03-2020
Add to basket
10060292 View Documents Merck Millipore
ZWBASE012
Base cleaning pouch £278.00
Pack of 12 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • For cleaning RO membrane in Elix™, Milli-RO™, Milli-RX™, and low-volume Millipore™ RiOs™ Water Purification Systems
  • Available in acidic (ROClean A) and basic (ROClean B) formulas
†For details, see Catalog 2004/05.
Specifications
Specifications
Elix™, Milli-RO™, Milli-RX™ and Millipore™ RiOs™ water purification system