Learn More
Merck Millipore RoClean™ Cleaning Pouches for Water Purification Systems
Keep your purification system clean with easy-to-use RoClean pouches.
Brand:  Merck Millipore ZWACID012
Additional Details : Weight : 0.50000kg Transport : UN number : 1727 Chem class : 8 Pack group : II
 View more versions of this product
Product Code. 10040272
  • £278.00 / Pack of 12
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • For cleaning RO membrane in Elix™, Milli-RO™, Milli-RX™, and low-volume Millipore™ RiOs™ Water Purification Systems
  • Available in acidic (ROClean A) and basic (ROClean B) formulas
Specifications
Specifications
Elix™, Milli-RO™, Milli-RX™ and Millipore™ RiOs™ water purification system
Acid cleaning pouch