missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Replacement Receiver Flasks

Provide a large filter area for rapid sterile filtration of aqueous fluids into narrow-mouth (33mm) or wide-mouth (45mm) bottles

Brand:  EMD Millipore™ SC00B02RE

Code : 71

Additional Details : Weight : 1.54724kg

 View more versions of this product

Product Code. 10156280

  • £46.25 / Pack of 12


In Stock 4
Add to basket

Description

Description

  • Sterile, polystyrene
  • Can be used as receivers for Millipore Steritop™ Vacuum Bottle-Top Filters
  • 45mm neck with screw-top lid

  • Storage of sterilized or clarified fluids
  • Laboratory Filtration

Specifications

Specifications

95mm
PS (Polystyrene)
Sterile
250mL
10.2cm
Videos
Documentation