Learn More
EMD Millipore™ Replacement Parts for Stirred Cells
Suitable for use with Amicon™ stirred cell apparatus
Brand:  EMD Millipore™ 8000SCPKIT
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11880512
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Specifications
Amicon™ 8000 series stirred cell apparatus
2 O-rings (2122S, 2118S, 2015S), tubing (4500202), tubing fitting assembly (15025AM) and membrane holder (2214801)
Replacement Part
Videos