Learn More
EMD Millipore™ Replacement Parts for Stirred Cells
Suitable for use with Amicon™ stirred cell apparatus
Brand:  EMD Millipore™ 8400SCPKIT
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10607515
  • £170.00 / 1 set
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications
Specifications
Amicon™ 8400 series stirred cell apparatus
Cap assembly (1524901), stirrer assembly (15247) and base (210000)
Replacement Part
Videos