Learn More
EMD Millipore™ Replacement Parts for Stirred Cells
Suitable for use with Amicon™ stirred cell apparatus
Brand:  EMD Millipore™ 8010SCPKIT
Code : 72
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11710403
  • £160.00 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Small Replacement Parts for 8000 Series Stirred Cells include 2 O-rings (Mfr. Nos. 2122S, 2118S, 2015S), tubing (Mfr. No. 4500202), tubing fitting assembly (Mfr. No. 15025AM, and membrane holder (Mfr. No. 2214801)
  • Large Replacement Parts for Stirred Cells include one each: cap assembly, stirrer assembly and base
Specifications
Specifications
Amicon™ 8010 series stirred cell apparatus
Cap assembly (1524901), stirrer assembly (15247) and base (210000)
Replacement Part
Videos