missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Replacement Parts for 90mm Glass Filter Holder

Replacement parts for 90mm All-glass filter holder

Brand:  EMD Millipore™ XX1009000

Code : 72

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10345141

  • £214.00 / Each


Estimated Shipment
12-06-2018
Add to basket

Specifications

Specifications

Funnel
Borosilicate Glass
90mm All-glass Filter Holder
1L
Videos
Documentation