missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Purospher™ STAR RP-18 Endcapped Hibar™ RT HPLC Columns, 3 micron

Fast method development of complex samples across the pH range from 1.5 to 10.5 at different mobile phases and temperature conditions

Brand:  EMD Millipore™ 1504310001

Code : Z3

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 15328163

  • £822.67 / Each


Estimated Shipment
04-06-2018
Add to basket

Description

Description

  • Highest silica purity (99.999%) for excellent peak symmetry
  • High separation efficiency
  • Reproducible results from run-to-run and from batch-to-batch
  • No phase collapse when using highly aqueous mobile phases
  • Good suitability for LC-MS applications
  • The Hibar RT columns are complete with endfittings

Specifications

Specifications

4mm
1/Pk.
HPLC
125mm
330m2/g
Videos