Learn More
Merck™ Purification Kit
With ion-exchange technology
Brand:  Merck™ QGARD00D3
Code : NEW
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10001592
  • £475.00 / Each
Estimated Shipment
09-12-2019
Description
Description
Q-Gard™ 3 Purification Cartridge for Milli-Q™ systems connected to DI water feed containing high silt
Specifications
Specifications
Q-Gard 3
Purification Cartridge
Depth Filtration, Ion-Exchange, Activated Carbon
Deionized
Milli-Q™systems connected to DI water feed containing high silt