Learn More
Merck PureProteome™ Nickel Magnetic Beads
Offer yields comparable with other magnetic purification systems while offering high purities
Brand: Merck LSKMAGH10
Code : X2
Additional Details : Weight : 0.01000kg
Description
- No lysate clearing—crude cell lysates are processed by magnetic beads
- Fast processing time—entire protocol is completed in less than one hour
- High capacity—bind 5 to 15mg of protein per mL of settled beads
- High affinity—binding can take place in buffer containing <5mM EDTA
- Flexible scale—varying the bead quantity allows different protein yields
- Flexible elution—native or denatured proteins
- Automatable—hands-free screening process
Specifications
1 to 5.5mg/mL | |
10 μm | |
Purification of His-tagged recombinant Protein |
2°C to 8°C | |
10 mL | |
Magnetic Affinity Bead |