Learn More
Merck Millipore PureProteome™ Magnetic Beads
For rapid and reproducible isolation of proteins
Brand: Merck Millipore LSKMAGN01
Code : NEW
Additional Details : Weight : 0.27000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Description
Custom affinity bead generation for binding to target ligand
- Advanced chemistry combined with high surface area provides more binding sites for proteins
- Particles visible as they adhere to side of tube for quick and easy aspiration and complete buffer removal
- Beads are immobilized in seconds
- Increased kinetics of bead-protein binding enables shorter incubations
- Significantly more affordable then competitive magnetic beads
Specifications
NHS flexiBind Magnetic bead system | |
Immunoprecipitations, purifying Nucleic Acids, isolating cells/organelles, Protein-protein interactions | |
2° to 8°C |
1 x 0.5mL | |
10μm |
Safety and Handling
- PureProteome NHS FlexiBind Magnetic Bead Kit
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
- MIXTURE LIST-Contains : 2-propanol