Learn More
Merck PureProteome™ Carboxy FlexiBind Magnetic Bead System
For rapid and reproducible isolation of proteins
Brand: Merck LSKMAG25CBX10
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Custom affinity bead generation for binding to target ligand.
- Advanced chemistry combined with high surface area provides more binding sites for proteins
- Particles visible as they adhere to side of tube for quick and easy aspiration and complete buffer removal
- Beads are immobilized in seconds
- Increased kinetics of bead-protein binding enables shorter incubations
- Significantly more affordable then competitive magnetic beads
Research Use Only
Specifications
Carboxy flexiBind Magnetic bead system | |
2.5μm | |
Immunoprecipitations, purifying Nucleic Acids, isolating cells/organelles, Protein-protein interactions |
10mL | |
2° to 8°C | |
Magnetic Beads |