missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Pure Water Storage Reservoir

Choose a Millipore Pure Water Storage Reservoir to store purified water without compromising quality.

Brand:  EMD Millipore™ TANKPE060

Code : Z1

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10105762

  • £952.95 / Each


Estimated Shipment
27-06-2018
Add to basket

Description

Description

Key Features

  • Polyethylene construction minimizes extractables that can contaminate water
  • Cylindrical shape minimizes area in contact with water
  • Smooth inner surface prevents biofilm formation
  • Conical bottom allows complete drainage for effective cleaning and sanitization
  • Hermetically sealed lid on large reservoir opening prevents air from leaking into reservoir and facilitates manual sanitization

Filling and Dispensing Features

  • Sensor-rod float triggers automatic reservoir refill
  • Water is fed into bottom of reservoir to prevent absorption of CO2
  • Two bottom valves allow direct connection to water polishing system and glassware washer
  • Water dispenses through valve on front of reservoir
Optional Features
  • Vent filter (available separately) guards against airborne impurities
  • 30, 60 and 100L reservoirs can be wall-mounted (require TANK FIX 01 wall mounting bracket)
  • Automatic Sanitization Module (A.S.M.) available separately to reduce bacteria levels
  • Optional distribution pump continuously delivers water under pressure
  • Water detector sensor (available separately) detects water on most surfaces such as floors and lab bench

Specifications

Specifications

Reservoir
60L
38cm
15.85 gal., 60L
38.1cm
83.3cm
Polyethylene
Videos