Learn More
Merck™ Pretreatment Packs
For use in EMD Millipore ELIX and RiOs™ Systems
Brand:  Merck™ PR0G0NP02
Code : NEW
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 15341432
  • £399.00 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
For optimal protection of Reverse Osmosis membranes
Specifications
Specifications
Pretreatment pack
Elix™, RiOs™
Activated Carbon, depth filtration
Videos