missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Plate with Fibronectin Coating

Pre-coated ECM multiwell plates

Brand:  EMD Millipore™ PIFB24P05

Code : 71

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11760373

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

Cell Attachment, Cell Culture, Cell Differentiation, Immunocytochemistry

Specifications

Specifications

24-Well Cell Culture Microplate
Cell attachment, cell culture, cell differentiation, immunocytochemistry
12.8cm
24
Fibronectin
1.9cm2
24 well
Polystyrene
Non-sterile
8.55cm
Videos