missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Petri Dishes and Petri-Pad™ Dishes

Available with or without absorbent pads

Brand:  EMD Millipore™ PD2004705

Code : NEW

Additional Details : Weight : 0.75000kg

 View more versions of this product

Product Code. 11983856

  • £169.33 / Pack of 600


Call for latest availability
+44 (0)1509 555 500
Add to basket

Requires : When using with liquid medium, add sterile absorbent pads (order Cat. No. AP10 047 S0, 47mm Sterile Pads; or AP10 045 S1, Millipore Pad Dispenser with Sterile Pads)

Description

Description

Petri Dishes

  • 47mm sterilized Petri dishes
  • Use with solid agar medium
  • Dishes are made of molded polystyrene
Petri-Pad Dishes
  • Presterilized disposable 47mm dishes preloaded with sterilized absorbent pads
  • Eliminate labor and cost of hand-loading pads and minimize contamination
  • Pads are pure cellulose

Specifications

Specifications

47 mm
600 Pk.
Yes
Dish without pad
Sleeve
Polystyrene
For solid agar medium or liquid medium with sterile absorbent pads
Yes
Videos