missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Petri Dishes and Petri-Pad™ Dishes

Available with or without absorbent pads

Brand:  EMD Millipore™ PD2004700

Code : 04

Additional Details : Weight : 1.36300kg

 View more versions of this product

Product Code. 11885863

  • £71.40 / Pack of 150


In Stock 3
Add to basket

Requires : When using with liquid medium, add sterile absorbent pads (order Cat. No. AP10 047 S0, 47mm Sterile Pads; or AP10 045 S1, Millipore Pad Dispenser with Sterile Pads)

Description

Description

Petri Dishes

  • 47mm sterilized Petri dishes
  • Use with solid agar medium
  • Dishes are made of molded polystyrene
Petri-Pad Dishes
  • Presterilized disposable 47mm dishes preloaded with sterilized absorbent pads
  • Eliminate labor and cost of hand-loading pads and minimize contamination
  • Pads are pure cellulose

Specifications

Specifications

47 mm
100 Pk. (5 sleeves); 20/Sleeve
Yes
Dish without pad
Sleeve
Polystyrene
For solid agar medium or liquid medium with sterile absorbent pads
Yes
Videos