missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Pellicon™ XL Devices

Small-volume tangential flow filter device

Brand:  EMD Millipore™ PXB100C50

Code : 72

Additional Details : Weight : 0.23000kg

 View more versions of this product

Product Code. 13111664

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

7.4 x 1.2 in. (18.8 x 3cm)
50cm2
Operating instructions and certificate of quality
Polypropylene
100kDa
0.03 to 0.05L/min.
15mL
Protein purification, analytical sample preparation
1 to 14
Ultrafiltration Biomax-100
Cassette
18.8cm
80psi at 50°C
100μm
3.2mL
3cm
Pellicon™ XL Ultrafiltration Device
1 Pack
Videos
Documentation

Technical Documents