missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Pellicon™ XL Devices

Small-volume tangential flow filter device

Brand:  EMD Millipore™ PXC030C50

Code : 72

Additional Details : Weight : 0.23000kg

 View more versions of this product

Product Code. 13197494

  • £560.00 / Each


Estimated Shipment
26-03-2018
Add to basket

Specifications

Specifications

7.4 x 1.2 in. (18.8 x 3cm)
50cm2
Operating instructions and certificate of quality
Polypropylene
30kDa
0.03 to 0.05L/min.
15mL
Protein purification, analytical sample preparation
2 to 21
Ultrafiltration Ultracel-30 PLCTK
Cassette
18.8cm
80psi at 50°C
100μm
3.2mL
3cm
Pellicon™ XL Ultrafiltration Device
1 Pack
Videos
Documentation

Technical Documents