Learn More
Merck Millipore Sterile Absorbent Pads
Dispensing of sterile absorbent pads while reducing contamination risks
Brand:  Merck Millipore AP10045S0
Code : 71
Additional Details : Weight : 0.70688kg
Product Code. 13188053
  • £258.71 / Pack of 1000
In stock, ready to ship: 2
Add to basket
Description
Description
  • Rapidly dispenses large numbers of absorbent pads without individual forceps handling
  • Plastic dispenser comes with tube that holds pads
  • Sterile pads are absorbent and conform with requirements of Standard Methods for the Examination of Water and Wastewater
Specifications
Specifications
47mm
Sterile Pads
Sterile
Pure Cellulose
1000 pads