Learn More
EMD Millipore™ Packs and Kits for Millipore Compact Water Systems For Milli-Q™ Compact Systems
Combination of purification media selected to remove typical contaminants
Brand:  EMD Millipore™ CPMQ004R1
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10532123
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
  • Replacement cartridges snap in and out
Specifications
Specifications
2
Reverse osmosis, distilled water
Ion exchange, activated Carbon
Quantum IX (Ionex)1
Milli-Q and Milli-RO Purification Systems
Videos