Learn More
Merck™ Packs and Kits for Compact Water Systems For Milli-Q™ Compact Systems
Combination of purification media selected to remove typical contaminants
Brand: Merck™ CPMQK05D2
Code : NEW
Additional Details : Weight : 0.10000kg
Description
- Replacement cartridges snap in and out
Specifications
QPAK 2 | |
Deionized water | |
Milli-Q and Milli-RO Purification Systems |
0.2μm | |
Purification pack, Millipak™ final filter | |
Ion exchange, depth filtration, membrane filter |