Learn More
Merck Millipore Nylon Woven Net Filters
Compatible with a broad range of solvents
£88.40 - £392.00
Specifications
Color Code White
Color (Overall) White
Product Type Net Filter
Feedwater Temperature (Metric) 100°C
Filter Type Plain
View More Specs
Products 18
Product Code Brand Pore Density Pore Size Price Quantity & Availability  
Product Code Brand Pore Density Pore Size Price Quantity & Availability  
10647321
View Documents
Merck Millipore
NY1H04700
44% 100μm £165.00
Pack of 100
Qty Price Break Available View alternative products
Save Up to 
»
Estimated Shipment
04-01-2021
Add to basket
10218400
View Documents
Merck Millipore
NY1104700
6% 11μm £138.00
Pack of 100
Qty Price Break Available View alternative products
Save Up to 
»
Estimated Shipment
14-12-2020
Add to basket
10258490
View Documents
Merck Millipore
NY1H02500
44% 100μm £105.00
Pack of 100
Qty Price Break Available View alternative products
Save Up to 
»
Estimated Shipment
14-12-2020
Add to basket
10703141
View Documents
Merck Millipore
NY2002500
14% 20μm
£88.40
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment
17-12-2020
Add to basket
10723121
View Documents
Merck Millipore
NY2004700
14% 20μm £173.00
Pack of 100
Qty Price Break Available View alternative products
Save Up to 
»
Estimated Shipment
15-12-2020
Add to basket
10339510
View Documents
Merck Millipore
NY2H04700
49% 120μm
£113.00
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment
14-01-2021
Add to basket
10773121
View Documents
Merck Millipore
NY4102500
31% 41μm
£88.40
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment
22-01-2021
Add to basket
10752151
View Documents
Merck Millipore
NY3002500
17% 30μm
£88.40
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment
04-01-2021
Add to basket
10084600
View Documents
Merck Millipore
NY3004700
17% 30μm £126.00
Pack of 100
Qty Price Break Available View alternative products
Save Up to 
»
Estimated Shipment
14-12-2020
Add to basket
10196530
View Documents
Merck Millipore
NY4104700
31% 41μm £175.00
Pack of 100
Qty Price Break Available View alternative products
Save Up to 
»
Estimated Shipment
21-12-2020
Add to basket
10556912
View Documents
Merck Millipore
NY6004700
41% 60μm
£120.00
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment
24-12-2020
Add to basket
10741991
View Documents
Merck Millipore
NY4H02500
43% 140μm
£96.44
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment
21-12-2020
Add to basket
10074660
View Documents
Merck Millipore
NY6H04700
52% 160μm
£113.00
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment
14-01-2021
Add to basket
10257820
View Documents
Merck Millipore
NY8002500
40% 80μm
£93.68
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment
21-12-2020
Add to basket
10014690
View Documents
Merck Millipore
NY8004700
40% 80μm
£113.00
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment
23-12-2020
Add to basket
10449840
View Documents
Merck Millipore
NY8H04700
47% 180μm £120.00
Pack of 100
Qty Price Break Available View alternative products
Save Up to 
»
Estimated Shipment
14-01-2021
Add to basket
10156440
View Documents
Merck Millipore
NY1H00010
44% 100μm
£392.00
Each
View alternative products
Save Up to 
»
Estimated Shipment
14-01-2021
Add to basket
10238250
View Documents
Merck Millipore
NY2H02500
49% 120μm
£88.40
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment
22-12-2020
Add to basket
Description
Description
  • Hydrophilic
  • Plain surface
  • Sterilization: Gamma or EO compatible
  • Operating Temperature: 75°C max.
  • Color: white
  • Mesh openings range from 10 to 180μm
  • Ideal for collection of algae and cells, particle analysis, large particulate filtration, background filter for automated particle imaging systems, prefiltration of solvents, paint monitoring
Specifications
Specifications
White
Net Filter
Plain
Hydrophilic
100°C
White
100°C
Particle filtration, particle analysis, prefiltration of solvents and paint monitoring
Nylon
SDS