Learn More
Merck Steriflip™ Sterile Centrifuge Tube Top Filter Units
Sterile, disposable filter unit for cell isolation.
Brand: Merck SCNY00040
Code : X2
Additional Details : Weight : 0.09450kg
Description
OverviewThe Steriflip filter unit is designed to work with standard 50 mL centrifuge tubes, which eliminates sample transfer. To filter, just attach the unit to a 50 mL centrifuge tube containing your sample, flip it over, and apply the vacuum. When finished, the filtrate collects in the attached 50 mL centrifuge tube.
New pore sizes ideal for Cell Isolation applications
Steriflip Nylon Net filters are now available in 40, 60 and 100 μm pore sizes, making them ideal for stem cell, cardiomyocite, neurological and skeletal tissue applications. The vacuum-assisted, closed system option ensures sample sterility while enabling faster separation of large volumes of cellular material and improved recoveries.
The Steriflip™ filter unit eliminates messy pouring during sample transfer. Directly attach the unit to a 50 mL centrifuge tube containing your sample, flip it over, and apply vacuum. The filtrate collects in the attached 50 mL centrifuge tube. Steriflip™ filters come in different pore sizes that are ideal for cell isolation. These include nylon net filters in 40, 60 and 100 μm pore sizes, which are ideal for stem cell, cardiomyocyte, neurological and skeletal tissue applications. The vacuum-assisted, closed system ensures sample sterility while enabling fast separation of large volumes of cellular material for efficient recoveries.
Features & Benefits
- Eliminates messy sample transfer via a simple vacuum-assisted, closed system
- Isolate cells from cellular debris using any of the various pore-sized nylon net filters
- Ideal for sterilizing small batches of culture media
- Attaches to any standard 50 mL centrifuge tube, eliminating unnecesary materials, time and risk of spillage
Applications: Sterilization of Aqueous Solutions (up to 50 mL), Sterile Filtration of Small Batches of Culture Media, Sterile Filtration of Buffers with High Value Protein or Small Molecule Components, Cell Isolation
Specifications
Clear | |
Single-use | |
Nylon Net | |
Sterile | |
Removal of coarse particulates, laboratory filtration | |
25 Pack | |
50 mL | |
5.43 in. | |
6.4 cm |
Double lead thread with vacuum port | |
MBS/Polypropylene | |
40 μm | |
7 cm2 | |
25Pack | |
Steriflip-NY Filter Unit | |
50 mL | |
13.8 cm |