Learn More
EMD Millipore™ Nutrient Media
Developed specifically for culturing microorganisms on membrane filters; select media type to match organism type and analytical test needs
Brand:  EMD Millipore™ MHA000P2M
Code : 72
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 13164624
  • £86.67 / 50 tests
Estimated Shipment
13-06-2018
Add to basket
Description
Description
  • Each dry powder lot for media preparation assayed for purity and compared to satisfactory lot
  • Each m-Endo broth for total coliform lot and m-FC broth for fecal coliform analysis lot certified in accordance with the Safe Drinking Water Act
  • Lot QC certificates included in each package as added assurance for critical procedures
Specifications
Specifications
Green
Liquid
M-Green Selective Broth
50 x 2mL
Testing soft drinks and other beverages for growth of Yeast and mold
Yeast and mold
United States
Ampule
2-10°C
Videos