missing translation for 'onlineSavingsMsg'
Learn More

Avantor™ J.T. Baker™ Filtration Columns with Frits, Bakerbond spe™

Empty filtration SPE columns packed with dual 20μm frits. For use with spe-12G and spe-24G glass column processors.

Brand:  Avantor™ 7121-03

250.80 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.



Code : NEW

Additional Details : Weight : 0.19000kg

Product Code. 10686631

  • £264.00 / Pack of 50
Estimated Shipment: 01-10-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

3 mL
Solid Phase Extraction Column
Pk. of 50
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers