missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ Multiwell Plate with Collagen I Coating

Enables most adherent cells to attach and proliferate

Brand:  EMD Millipore™ PICL06P05

Code : 71

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11710373

  • £74.67 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

6- and 24-well format polystyrene plates provide flexibility to run multiple samples simultaneously

  • Uses ECM proteins combined with cellular adhesion molecules on the surface of cells
  • Facilitates necessary substrate for many fastidious cells to attach to the surface
  • Allows efficient plating and higher proliferation rate for cells
  • Produces demonstrable enhanced differentiation potential in cells
  • Non-sterile

Order Info

Individually blister packaged in packs of five

Specifications

Specifications

6-Well Multiwell Plate
Cell attachment, cell culture, cell differentiation, immunocytochemistry
12.8cm
6
Collagen I
9.6cm2
6 well
Polystyrene
Non-sterile
8.55cm
Videos