Learn More
Merck MultiScreenHTS Solubility Filter Plate
96-well filter plate designed and optimized for aqueous solubility assays
Brand: Merck MSSLBPC50
Code : Z1
Additional Details : Weight : 0.10000kg
Description
- High-throughput system to classify or quantify the aqueous solubility of compound libraries stored in solvent as a concentrated solution
- Analysis time in under four hours using <100μg of sample per test
- Automation compatible, with space for bar coding
- Validated for high drug recovery Low-binding membrane and low-binding plate materials
Specifications
Standard | |
8 x 12 | |
Clear | |
Styrene | |
Non-sterile | |
50/Pk. |
Polycarbonate | |
0.4 μm | |
MultiScreenHTS-PCF Filter Plate | |
96 | |
Aqueous solubility assays |