missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ MultiScreen™HTS 96-Well Filter Plates with Negatively Charged Membrane

Binds positive charge—ideal for kinase assays with peptide substrates and enzyme assays

Brand:  EMD Millipore™ MSPHNXB50

Code : 73

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 10347613

  • £986.19 / Pack of 50


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

MultiScreenHTS+ Hi Flow PH Plate
Opaque
0.26cm2
1.28cm
Negatively Charged Phosphocellulose, Polyester Mesh
0.65μm
0.05 to 0.25mL
8.55cm
ANSI, SBS
0.5 x 3.36 x 0.56 in. (1.28 x 8.55 x 1.44cm)
Protein kinase assays with peptide substrates, enzyme assays, binds positive charge
Barex&trade
96
Non-sterile
1.44cm
Videos