missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreenHTS 96-Well Filter Plates
For filtration-based solubility determination. MultiScreenHTS 96-Well Filter Plates are specifically developed for use with automation
Brand: Merck MSFBN6B50
Code : 73
Additional Details : Weight : 4.32000kg
Description
- High Throughput
- Meet ANSI/SBS 2004 plate specifications
- Protects underdrain, eliminates surface contact with individual well ‘drip directors’
- Plastic skirt enables stacking directly with collection plates
Applications
- Receptor-ligand binding; general sample preparation; Elispot; enzyme activity; bead-based assays; whole organism visualisation
- Radioisotope detection
Specifications
ANSI, SBS | |
Hydrophilic PVDF | |
1/0.65 μm | |
Barex | |
Non-sterile | |
Receptor, ligand binding assays, sample prep, enzyme assays with precipitation, DNA purification | |
0.5 x 8.55 x 0.56 in. (1.28 x 21.71 x 1.44 cm) | |
21.71 cm |
Standard | |
8 x 12 | |
Opaque | |
96 | |
0.26 cm2 | |
50/Pk. | |
1.28 cm | |
1.44 cm |