missing translation for 'onlineSavingsMsg'
Learn More

EMD Millipore™ MultiScreenHTS Durapore™ 96-Well Filter Plates

Designed for full automation compatibility. EMD Millipore MultiScreenHTS Durapore™ 96-Well Filter Plates are innovative plates ideal for bead based assays. Opaque Barex™ and clear, sterile or non-sterile styrene and racks are available in a range of pore sizes.

Brand:  EMD Millipore™ MSGVN2210

Code : 73

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10367123

  • £249.00 / Pack of 10


Estimated Shipment
12-07-2018
Add to basket

Description

Description

For use in receptor-ligand binding, general sample preparation, ELISPOT, enzyme activity, bead-based assays, whole organism visualization, and radioisotope detection.
  • Standardized dimensions and aligned, rigid sidewalls
    • Allows full compatibility with automated/robotic gripper arms, stackers, plate washers, bar code labels, and plate readers
  • Unique two-piece plate design eliminates surface contact with individual well “drip directors” to prevent crosstalk between wells
  • Extended skirt and vacuum manifolds
    • Allows easy, contamination-free stacking directly with collection plates
    • Protects recessed underdrain
    • Provides ample surface for bar code labels
    • Improves vacuum seal
  • Removable underdrain allows membrane access for ELISPOT assays, whole organism visualization, and radioisotope detection
  • Isolated well design and individually sealed filter discs eliminate potential for reagent flow from well to well
  • Opaque plastic prevents light transfer between wells during direct scintillation counting
  • Low binding hydrophilic Durapore™ membrane allows low nonspecific binding of antibodies, reagents, and samples
  • Meet ANSI™/SBS 2004 plate specifications
Specifications

Specifications

MultiScreenHTS-GV Plate
ANSI, SBS
0.26cm2
Styrene
96
Pk. of 10
0.05 to 0.25mL
Low binding
Clear
Receptor binding assays on whole cells and cell fragments, protein kinase, phosphatase precipitation assays, bead, resin based assays
Hydrophilic PVDF
0.22μm
Non-sterile
Videos
Documentation

Technical Documents

View accessory products