missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreenHTS 96-Well Filter Plates
For filtration-based solubility determination. MultiScreenHTS 96-Well Filter Plates are specifically developed for use with automation
Brand: Merck MSFBN6B10
Code : 73
Additional Details : Weight : 0.80000kg
Description
- High Throughput
- Meet ANSI/SBS 2004 plate specifications
- Protects underdrain, eliminates surface contact with individual well ‘drip directors’
- Plastic skirt enables stacking directly with collection plates
Applications
- Receptor-ligand binding; general sample preparation; Elispot; enzyme activity; bead-based assays; whole organism visualisation
- Radioisotope detection
Specifications
ANSI, SBS | |
Hydrophilic PVDF | |
1/0.65 μm | |
Opaque | |
96 | |
0.26 cm2 | |
10/Pk. |
Standard | |
8 x 12 | |
Low Binding | |
Barex | |
Non-sterile | |
Receptor, ligand binding assays, sample prep, enzyme assays with precipitation, DNA purification |