Learn More
EMD Millipore™ MultiScreenHTS Durapore™ 96-Well Filter Plates
Designed for full automation compatibility. EMD Millipore MultiScreenHTS Durapore™ 96-Well Filter Plates are innovative plates ideal for bead based assays. Opaque Barex™ and clear, sterile or non-sterile styrene and racks are available in a range of pore sizes.
Brand: EMD Millipore™ MSDVN6510
Code : 73
Additional Details : Weight : 0.68432kg
Description
For use in receptor-ligand binding, general sample preparation, ELISPOT, enzyme activity, bead-based assays, whole organism visualization, and radioisotope detection.- Standardized dimensions and aligned, rigid sidewalls
- Allows full compatibility with automated/robotic gripper arms, stackers, plate washers, bar code labels, and plate readers
- Unique two-piece plate design eliminates surface contact with individual well “drip directors” to prevent crosstalk between wells
- Extended skirt and vacuum manifolds
- Allows easy, contamination-free stacking directly with collection plates
- Protects recessed underdrain
- Provides ample surface for bar code labels
- Improves vacuum seal
- Removable underdrain allows membrane access for ELISPOT assays, whole organism visualization, and radioisotope detection
- Isolated well design and individually sealed filter discs eliminate potential for reagent flow from well to well
- Opaque plastic prevents light transfer between wells during direct scintillation counting
- Low binding hydrophilic Durapore™ membrane allows low nonspecific binding of antibodies, reagents, and samples
- Meet ANSI™/SBS 2004 plate specifications
Specifications
MultiScreenHTS-DV Plate | |
ANSI, SBS | |
0.28cm2 | |
Styrene | |
96 | |
Pk. of 10 | |
0.05 to 0.25mL |
Low binding | |
Clear | |
Receptor binding assays on whole cells and cell fragments, protein kinase, phosphatase precipitation assays, bead, resin based assays | |
Hydrophilic PVDF | |
0.65μm | |
Non-sterile |