Learn More
Merck MultiScreen™ Solvinert Filter Plates
Optimized for drug discovery applications. MultiScreen™ Solvinert Filter Plates are solvent compatible and feature a unique one-piece design. 96-well plates are available in deep well (2mL) format (with or without pre-filter) and standard (500μL) format, with hydrophobic or hydrophilic membranes.
Brand: Merck MDRLN0410
Code : 73
Additional Details : Weight : 0.10000kg
Description
- Clear polyolefin base plate offers broad solvent compatibility
- One-piece design eliminates leaking and allows for extended incubations
- Low binding and low extractables
- Accommodate aqueous and nonaqueous samples with chemically resistant hydrophilic or hydrophobic membranes
- Polytetrafluoroethylene membranes feature 45μL pores
- Automation compatible
- Available in 96-well deep well (2mL) format (with or without pre-filter) and standard (500μL) format
Specifications
Standard | |
8 x 12 | |
Clear | |
96 | |
0.28cm2 | |
10/Pk. | |
1.28 cm | |
4.07 cm |
Hydrophilic PTFE | |
0.45 μm | |
Polyolefin Copolymer | |
Non-sterile | |
Solid phase synthesis and cleanup, in plate protein precipitation, sample recovery following in-plate compound cleavage, peptide synthesis | |
10/Pk. | |
8.55 cm |